Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013011.3 | NP_037143.2 | 2 - 3 | 354 | 104 | |
XM_006241529.2 | XP_006241591.1 | 3 - 4 | 907 | 104 | ||
GenBank mRNA | BC070941.1 | - | 2 - 3 | 354 | 7099670 | 104 |
BC094305.1 | - | 2 - 3 | 659 | 7313193 | 104 | |
D17615.1 | - | 2 - 3 | 421 | 104 | ||
D30740.1 | - | 2 - 3 | 334 | 104 | ||
U37252.1 | - | 2 - 3 | 355 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Endogenous Controls Recommended Assay Rattus norvegicus Rat Chemokine Panel Rat Endogenous Controls Inventoried Rat_Endogenous_Controls Probe spans exons |