Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013166.1 | NP_037298.1 | 1 - 2 | 193 | 83 | |
GenBank mRNA | FQ216326.1 | - | 1 - 2 | 156 | 83 | |
X17457.1 | - | 1 - 2 | 193 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |