Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022293.1 | NP_071629.1 | 1 - 2 | 427 | 76 | |
XM_006240447.2 | XP_006240509.1 | 2 - 3 | 3225 | 76 | ||
GenBank mRNA | AF287301.1 | - | 1 - 2 | 427 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |