Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138883.1 | NP_620238.1 | 6 - 7 | 528 | 114 | |
GenBank mRNA | BC060544.1 | - | 6 - 7 | 536 | 6918775 | 114 |
D13127.1 | - | 6 - 7 | 528 | 114 | ||
FQ209940.1 | - | 6 - 7 | 536 | 114 | ||
FQ210807.1 | - | 6 - 7 | 542 | 114 | ||
FQ217675.1 | - | 6 - 7 | 533 | 114 | ||
FQ223905.1 | - | 6 - 7 | 565 | 114 | ||
FQ226488.1 | - | 6 - 7 | 529 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |