Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182814.2 | NP_877966.1 | 10 - 11 | 1251 | 71 | |
GenBank mRNA | AY223861.1 | - | 10 - 11 | 1117 | 71 | |
BC079283.1 | - | 10 - 11 | 1222 | 7134210 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |