Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182825.1 | NP_877977.1 | 1 - 2 | 239 | 86 | |
XM_008765754.1 | XP_008763976.1 | 1 - 2 | 257 | 86 | ||
XM_008765755.1 | XP_008763977.1 | 1 - 2 | 257 | 86 | ||
GenBank mRNA | AY327411.1 | - | 1 - 2 | 239 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |