Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182844.1 | NP_878264.1 | 2 - 3 | 333 | 73 | |
XM_006244110.2 | XP_006244172.1 | 2 - 3 | 373 | 73 | ||
GenBank mRNA | AY331984.1 | - | 2 - 3 | 333 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |