Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182952.2 | NP_891997.1 | 2 - 3 | 255 | 67 | |
GenBank mRNA | AF537209.1 | - | 2 - 3 | 255 | 67 | |
AY340181.1 | - | 2 - 3 | 234 | 67 | ||
BC168725.1 | - | 2 - 3 | 263 | 6919932 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |