Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhal6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183334.1 | NP_899163.1 | 1 | 1364 | 68 | |
GenBank mRNA | AY342428.1 | - | 1 | 1364 | 68 | |
BC078970.1 | - | 1 | 1345 | 7111795 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |