Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012219.1 | NP_001012219.1 | 4 - 5 | 644 | 64 | |
XM_006233528.2 | XP_006233590.1 | 5 - 6 | 910 | 64 | ||
GenBank mRNA | BC090011.1 | - | 4 - 5 | 644 | 7324565 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |