Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013006.1 | NP_037138.1 | 5 - 6 | 355 | 128 | |
XM_008763522.1 | XP_008761744.1 | 4 - 5 | 347 | 128 | ||
GenBank mRNA | BC085750.1 | - | 5 - 6 | 293 | 7190638 | 128 |
D63885.1 | - | 5 - 6 | 355 | 128 | ||
U97146.1 | - | 5 - 6 | 383 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |