Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013096.1 | NP_037228.1 | 1 - 2 | 125 | 129 | |
GenBank mRNA | BC059150.1 | - | 1 - 2 | 124 | 6918464 | 129 |
BC091567.1 | - | 112 | 7458326 | 129 | ||
BC160818.1 | - | 1 - 2 | 108 | 7375173 | 129 | |
FQ209733.1 | - | 1 - 2 | 125 | 129 | ||
FQ209744.1 | - | 1 - 2 | 125 | 129 | ||
FQ209909.1 | - | 1 - 2 | 125 | 129 | ||
FQ209912.1 | - | 1 - 2 | 129 | |||
FQ210001.1 | - | 1 - 2 | 125 | 129 | ||
FQ210150.1 | - | 1 - 2 | 123 | 129 | ||
FQ210186.1 | - | 1 - 2 | 123 | 129 | ||
FQ210277.1 | - | 1 - 2 | 123 | 129 | ||
FQ210349.1 | - | 1 - 2 | 129 | |||
FQ210635.1 | - | 1 - 2 | 125 | 129 | ||
FQ210643.1 | - | 1 - 2 | 123 | 129 | ||
FQ210690.1 | - | 1 - 2 | 124 | 129 | ||
FQ210723.1 | - | 1 - 2 | 123 | 129 | ||
FQ210862.1 | - | 1 - 2 | 123 | 129 | ||
FQ210866.1 | - | 1 - 2 | 123 | 129 | ||
FQ211077.1 | - | 1 - 2 | 125 | 129 | ||
FQ211217.1 | - | 1 - 2 | 123 | 129 | ||
FQ211220.1 | - | 1 - 2 | 125 | 129 | ||
FQ211232.1 | - | 1 - 2 | 124 | 129 | ||
FQ211239.1 | - | 1 - 2 | 123 | 129 | ||
FQ211374.1 | - | 1 - 2 | 123 | 129 | ||
FQ211431.1 | - | 1 - 2 | 123 | 129 | ||
FQ211513.1 | - | 1 - 2 | 123 | 129 | ||
FQ211755.1 | - | 1 - 2 | 123 | 129 | ||
FQ211911.1 | - | 1 - 2 | 123 | 129 | ||
FQ211972.1 | - | 1 - 2 | 123 | 129 | ||
FQ212025.1 | - | 1 - 2 | 125 | 129 | ||
FQ212153.1 | - | 1 - 2 | 125 | 129 | ||
FQ212186.1 | - | 1 - 2 | 123 | 129 | ||
FQ212232.1 | - | 1 - 2 | 125 | 129 | ||
FQ212457.1 | - | 1 - 2 | 123 | 129 | ||
FQ212854.1 | - | 1 - 2 | 123 | 129 | ||
FQ213301.1 | - | 1 - 2 | 123 | 129 | ||
FQ214066.1 | - | 1 - 2 | 123 | 129 | ||
FQ214223.1 | - | 1 - 2 | 123 | 129 | ||
FQ216243.1 | - | 1 - 2 | 123 | 129 | ||
FQ217723.1 | - | 1 - 2 | 123 | 129 | ||
FQ217724.1 | - | 1 - 2 | 123 | 128 | ||
FQ217869.1 | - | 1 - 2 | 123 | 129 | ||
FQ217988.1 | - | 1 - 2 | 125 | 129 | ||
FQ218117.1 | - | 1 - 2 | 123 | 129 | ||
FQ218146.1 | - | 123 | 129 | |||
FQ218612.1 | - | 1 - 2 | 122 | 129 | ||
FQ218968.1 | - | 1 - 2 | 125 | 129 | ||
FQ219715.1 | - | 1 - 2 | 123 | 129 | ||
FQ220786.1 | - | 1 - 2 | 123 | 129 | ||
FQ221656.1 | - | 1 - 2 | 124 | 129 | ||
FQ221912.1 | - | 1 - 2 | 125 | 129 | ||
FQ221952.1 | - | 1 - 2 | 123 | 129 | ||
FQ222431.1 | - | 1 - 2 | 123 | 129 | ||
FQ222454.1 | - | 1 - 2 | 123 | 129 | ||
FQ222672.1 | - | 1 - 2 | 123 | 129 | ||
FQ222750.1 | - | 1 - 2 | 125 | 129 | ||
FQ223017.1 | - | 1 - 2 | 124 | 129 | ||
FQ224581.1 | - | 1 - 2 | 123 | 129 | ||
FQ225427.1 | - | 1 - 2 | 124 | 129 | ||
FQ225440.1 | - | 1 - 2 | 124 | 129 | ||
FQ225452.1 | - | 1 - 2 | 123 | 129 | ||
FQ225777.1 | - | 1 - 2 | 123 | 129 | ||
FQ225808.1 | - | 1 - 2 | 126 | 129 | ||
FQ226118.1 | - | 1 - 2 | 125 | 129 | ||
FQ226173.1 | - | 1 - 2 | 125 | 129 | ||
FQ226175.1 | - | 1 - 2 | 123 | 129 | ||
FQ226203.1 | - | 1 - 2 | 123 | 129 | ||
FQ226615.1 | - | 1 - 2 | 123 | 129 | ||
FQ227082.1 | - | 1 - 2 | 123 | 129 | ||
FQ228301.1 | - | 1 - 2 | 123 | 129 | ||
FQ228436.1 | - | 123 | 129 | |||
FQ230483.1 | - | 1 - 2 | 125 | 129 | ||
FQ230901.1 | - | 1 - 2 | 123 | 129 | ||
FQ231201.1 | - | 1 - 2 | 115 | 129 | ||
FQ231290.1 | - | 1 - 2 | 123 | 129 | ||
FQ232272.1 | - | 1 - 2 | 123 | 129 | ||
FQ232481.1 | - | 1 - 2 | 123 | 129 | ||
FQ234683.1 | - | 1 - 2 | 123 | 129 | ||
M17083.1 | - | 1 - 2 | 125 | 129 |
Gene Symbol: Hba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007722.1 | NP_001007723.1 | 125 | 129 | ||
GenBank mRNA | FQ209950.1 | - | 1 - 2 | 123 | 129 | |
FQ210108.1 | - | 1 - 2 | 123 | 129 | ||
FQ210256.1 | - | 1 - 2 | 123 | 129 | ||
FQ210264.1 | - | 1 - 2 | 123 | 129 | ||
FQ210372.1 | - | 1 - 2 | 128 | 129 | ||
FQ210420.1 | - | 1 - 2 | 123 | 129 | ||
FQ210570.1 | - | 1 - 2 | 123 | 129 | ||
FQ210694.1 | - | 1 - 2 | 123 | 129 | ||
FQ211180.1 | - | 1 - 2 | 123 | 129 | ||
FQ211270.1 | - | 1 - 2 | 123 | 129 | ||
FQ211366.1 | - | 1 - 2 | 123 | 129 | ||
FQ211672.1 | - | 1 - 2 | 123 | 129 | ||
FQ211887.1 | - | 1 - 2 | 123 | 129 | ||
FQ213126.1 | - | 1 - 2 | 123 | 129 | ||
FQ214258.1 | - | 1 - 2 | 123 | 129 | ||
FQ215087.1 | - | 1 - 2 | 123 | 129 | ||
FQ217874.1 | - | 1 - 2 | 123 | 129 | ||
FQ218140.1 | - | 1 - 2 | 125 | 129 | ||
FQ218297.1 | - | 1 - 2 | 129 | 129 | ||
FQ221489.1 | - | 1 - 2 | 123 | 129 | ||
FQ222342.1 | - | 1 - 2 | 123 | 129 | ||
FQ222639.1 | - | 1 - 2 | 122 | 129 | ||
FQ222693.1 | - | 1 - 2 | 121 | 129 | ||
FQ223754.1 | - | 1 - 2 | 125 | 129 | ||
FQ223838.1 | - | 1 - 2 | 149 | 129 | ||
FQ224218.1 | - | 1 - 2 | 132 | 129 | ||
FQ224339.1 | - | 1 - 2 | 125 | 129 | ||
FQ224679.1 | - | 1 - 2 | 123 | 129 | ||
FQ225311.1 | - | 1 - 2 | 123 | 129 | ||
FQ225451.1 | - | 1 - 2 | 123 | 129 | ||
FQ225711.1 | - | 1 - 2 | 123 | 129 | ||
FQ225789.1 | - | 1 - 2 | 125 | 129 | ||
FQ226185.1 | - | 1 - 2 | 123 | 129 | ||
FQ226226.1 | - | 278 | 129 | |||
FQ226238.1 | - | 1 - 2 | 113 | 129 | ||
FQ226556.1 | - | 1 - 2 | 123 | 129 | ||
FQ226574.1 | - | 1 - 2 | 123 | 129 | ||
FQ226576.1 | - | 1 - 2 | 123 | 129 | ||
FQ226594.1 | - | 1 - 2 | 123 | 129 | ||
FQ226619.1 | - | 1 - 2 | 123 | 129 | ||
FQ226927.1 | - | 1 - 2 | 123 | 129 | ||
FQ228017.1 | - | 1 - 2 | 125 | 129 | ||
FQ228292.1 | - | 1 - 2 | 123 | 129 | ||
FQ228298.1 | - | 1 - 2 | 123 | 129 | ||
FQ228306.1 | - | 1 - 2 | 123 | 129 | ||
FQ228588.1 | - | 1 - 2 | 123 | 129 | ||
FQ229556.1 | - | 1 - 2 | 123 | 129 | ||
FQ230501.1 | - | 1 - 2 | 123 | 129 | ||
FQ230536.1 | - | 1 - 2 | 123 | 129 | ||
FQ230819.1 | - | 1 - 2 | 125 | 129 | ||
FQ230868.1 | - | 1 - 2 | 123 | 129 | ||
FQ230877.1 | - | 1 - 2 | 125 | 129 | ||
FQ231222.1 | - | 1 - 2 | 123 | 129 | ||
FQ231239.1 | - | 1 - 2 | 118 | 129 | ||
FQ231246.1 | - | 1 - 2 | 123 | 129 | ||
FQ231250.1 | - | 1 - 2 | 123 | 129 | ||
FQ231268.1 | - | 1 - 2 | 122 | 129 | ||
FQ231572.1 | - | 1 - 2 | 123 | 129 | ||
FQ231902.1 | - | 1 - 2 | 123 | 129 | ||
FQ232213.1 | - | 1 - 2 | 123 | 129 | ||
FQ234805.1 | - | 1 - 2 | 123 | 129 | ||
FQ235117.1 | - | 1 - 2 | 124 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |