Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013226.2 | NP_037358.1 | 4 | 469 | 72 | |
GenBank mRNA | BC061562.1 | - | 4 | 469 | 6921029 | 72 |
BC082797.1 | - | 4 | 454 | 7301263 | 72 | |
FQ209939.1 | - | 4 | 465 | 72 | ||
FQ210972.1 | - | 4 | 462 | 72 | ||
FQ211501.1 | - | 4 | 462 | 72 | ||
FQ211742.1 | - | 4 | 466 | 72 | ||
FQ212686.1 | - | 4 | 462 | 72 | ||
FQ216988.1 | - | 4 | 465 | 72 | ||
FQ217782.1 | - | 4 | 465 | 72 | ||
FQ217854.1 | - | 4 | 465 | 72 | ||
FQ219795.1 | - | 4 | 462 | 72 | ||
FQ220681.1 | - | 4 | 463 | 72 | ||
FQ221082.1 | - | 4 | 465 | 72 | ||
FQ221405.1 | - | 3 | 431 | 72 | ||
FQ221557.1 | - | 4 | 461 | 72 | ||
FQ221558.1 | - | 4 | 466 | 72 | ||
FQ221666.1 | - | 4 | 465 | 72 | ||
FQ221830.1 | - | 4 | 466 | 72 | ||
FQ221978.1 | - | 4 | 466 | 72 | ||
FQ221986.1 | - | 4 | 465 | 72 | ||
FQ221995.1 | - | 4 | 466 | 72 | ||
FQ222044.1 | - | 4 | 462 | 72 | ||
FQ222154.1 | - | 4 | 464 | 72 | ||
FQ222265.1 | - | 4 | 465 | 72 | ||
FQ222417.1 | - | 4 | 462 | 72 | ||
FQ222458.1 | - | 4 | 466 | 72 | ||
FQ222536.1 | - | 4 | 465 | 72 | ||
FQ222629.1 | - | 4 | 462 | 72 | ||
FQ222759.1 | - | 4 | 466 | 72 | ||
FQ222997.1 | - | 4 | 466 | 72 | ||
FQ223042.1 | - | 4 | 465 | 72 | ||
FQ223180.1 | - | 4 | 465 | 72 | ||
FQ223244.1 | - | 4 | 462 | 72 | ||
FQ224329.1 | - | 4 | 462 | 72 | ||
FQ224364.1 | - | 4 | 466 | 72 | ||
FQ224436.1 | - | 4 | 462 | 72 | ||
FQ228463.1 | - | 4 | 465 | 72 | ||
FQ228512.1 | - | 4 | 462 | 72 | ||
FQ228892.1 | - | 4 | 469 | 72 | ||
X06483.1 | - | 3 | 420 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |