Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017151.2 | NP_058847.1 | 2 - 3 | 107 | 91 | |
GenBank mRNA | AA892895.1 | - | 2 - 3 | 91 | ||
FQ211784.1 | - | 2 - 3 | 107 | 91 | ||
FQ217117.1 | - | 2 - 3 | 108 | 91 | ||
FQ217914.1 | - | 2 - 3 | 107 | 91 | ||
FQ220999.1 | - | 2 - 3 | 107 | 91 | ||
FQ221086.1 | - | 2 - 3 | 108 | 91 | ||
FQ221189.1 | - | 2 - 3 | 107 | 91 | ||
FQ221217.1 | - | 2 - 3 | 108 | 91 | ||
FQ221800.1 | - | 2 - 3 | 107 | 91 | ||
FQ222134.1 | - | 2 - 3 | 107 | 91 | ||
FQ222137.1 | - | 2 - 3 | 108 | 91 | ||
FQ222200.1 | - | 2 - 3 | 107 | 91 | ||
FQ222233.1 | - | 2 - 3 | 106 | 91 | ||
FQ222539.1 | - | 2 - 3 | 107 | 91 | ||
FQ222610.1 | - | 2 - 3 | 107 | 91 | ||
FQ222928.1 | - | 2 - 3 | 107 | 91 | ||
FQ223014.1 | - | 1 - 2 | 109 | 91 | ||
FQ223311.1 | - | 2 - 3 | 108 | 91 | ||
FQ223478.1 | - | 2 - 3 | 110 | 91 | ||
FQ223811.1 | - | 2 - 3 | 107 | 91 | ||
FQ223860.1 | - | 2 - 3 | 103 | 91 | ||
FQ224530.1 | - | 2 - 3 | 107 | 91 | ||
FQ228367.1 | - | 2 - 3 | 115 | 91 | ||
FQ228405.1 | - | 2 - 3 | 110 | 91 | ||
M19393.1 | - | 2 - 3 | 107 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |