Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017245.2 | NP_058941.1 | 7 - 8 | 1409 | 62 | |
GenBank mRNA | BC066661.1 | - | 7 - 8 | 1409 | 5623322 | 62 |
Y07504.1 | - | 7 - 8 | 1395 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |