Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022539.1 | NP_071984.1 | 4 - 5 | 675 | 69 | |
GenBank mRNA | BC070892.1 | - | 4 - 5 | 590 | 7099147 | 69 |
BC091150.1 | - | 4 - 5 | 689 | 7122560 | 69 | |
L10652.1 | - | 4 - 5 | 675 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |