Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024148.1 | NP_077062.1 | 3 - 4 | 348 | 104 | |
XM_006251913.1 | XP_006251975.1 | 3 - 4 | 389 | 104 | ||
GenBank mRNA | AF309114.1 | - | 2 - 3 | 264 | 104 | |
AF311054.1 | - | 2 - 3 | 226 | 104 | ||
BC078816.1 | - | 3 - 4 | 356 | 7127205 | 104 | |
D44495.1 | - | 3 - 4 | 348 | 104 | ||
FQ220521.1 | - | 3 - 4 | 350 | 104 | ||
L27076.1 | - | 3 - 4 | 332 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |