Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 7 - 8 | 1012 | 94 | |
GenBank mRNA | BC059125.1 | - | 7 - 8 | 940 | 6919326 | 94 |
FQ209697.1 | - | 7 - 8 | 905 | 94 | ||
FQ210400.1 | - | 7 - 8 | 904 | 94 | ||
FQ213475.1 | - | 7 - 8 | 936 | 94 | ||
FQ218913.1 | - | 7 - 8 | 907 | 94 | ||
FQ218933.1 | - | 7 - 8 | 906 | 94 | ||
FQ219092.1 | - | 7 - 8 | 906 | 94 | ||
FQ225110.1 | - | 7 - 8 | 974 | 94 | ||
M17300.1 | - | 6 - 7 | 882 | 94 | ||
M34477.1 | - | 7 - 8 | 1012 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |