Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763804.1 | XP_008762026.1 | 1 - 2 | 414 | 77 | |
GenBank mRNA | AF072439.1 | - | 145 | 77 | ||
CB727538.1 | - | 1 - 2 | 391 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Probe spans exons |