Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133544.1 | NP_598228.1 | 2 - 3 | 198 | 113 | |
GenBank mRNA | AJ271158.1 | - | 2 - 3 | 198 | 113 | |
FQ217274.1 | - | 3 - 4 | 205 | 113 | ||
FQ217509.1 | - | 3 - 4 | 258 | 113 | ||
FQ217783.1 | - | 3 - 4 | 257 | 113 | ||
FQ221212.1 | - | 3 - 4 | 256 | 113 | ||
FQ223588.1 | - | 3 - 4 | 253 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |