Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138548.2 | NP_612557.1 | 2 - 3 | 231 | 72 | |
GenBank mRNA | D13374.1 | - | 2 - 3 | 232 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons |