Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138911.3 | NP_620266.1 | 10 - 11 | 1307 | 60 | |
GenBank mRNA | BC061529.1 | - | 10 - 11 | 1308 | 6887497 | 60 |
FQ220758.1 | - | 7 - 8 | 829 | 60 | ||
FQ227512.1 | - | 1 - 2 | 58 | 60 | ||
Y15068.1 | - | 9 - 10 | 1242 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |