Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147135.2 | NP_671476.1 | 1 - 2 | 489 | 91 | |
GenBank mRNA | AB010154.1 | - | 1 - 2 | 201 | 91 | |
BC131843.1 | - | 1 - 2 | 489 | 8363395 | 91 |
Gene Symbol: LOC103690016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758862.1 | XP_008757084.1 | 1 - 2 | 537 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |