Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031523.1 | NP_113711.1 | 2 - 3 | 249 | 143 | |
GenBank mRNA | BC078784.1 | - | 2 - 3 | 231 | 7128287 | 143 |
J00758.1 | - | 2 - 3 | 243 | 143 | ||
M11563.1 | - | 2 - 3 | 249 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons |