Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031646.1 | NP_113834.1 | 2 - 3 | 207 | 89 | |
GenBank mRNA | AB030943.1 | - | 2 - 3 | 216 | 89 | |
AB042888.1 | - | 2 - 3 | 207 | 89 | ||
AF181551.1 | - | 2 - 3 | 210 | 89 | ||
EF595744.1 | - | 2 - 3 | 131 | 89 | ||
FQ226087.1 | - | 2 - 3 | 232 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |