Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014192.1 | NP_001014214.1 | 5 - 6 | 690 | 65 | |
GenBank mRNA | BC082025.1 | - | 5 - 6 | 690 | 7129968 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |