Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 5 - 6 | 673 | 137 | |
XM_006256359.2 | XP_006256421.1 | 5 - 6 | 724 | 137 | ||
XM_006256361.2 | XP_006256423.1 | 5 - 6 | 747 | 137 | ||
XM_006256362.2 | XP_006256424.1 | 5 - 6 | 639 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |