Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257671.2 | XP_006257733.1 | 6 - 7 | 1135 | 83 | |
XM_006257672.2 | XP_006257734.1 | 7 - 8 | 1071 | 83 | ||
XM_006257673.2 | XP_006257735.1 | 7 - 8 | 1094 | 83 | ||
XM_006257674.2 | XP_006257736.1 | 7 - 8 | 1088 | 83 | ||
XM_006257675.2 | XP_006257737.1 | 6 - 7 | 1044 | 83 | ||
XM_006257676.2 | XP_006257738.1 | 6 - 7 | 1014 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |