Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098727.1 | NP_001092197.1 | 2 - 3 | 646 | 102 | |
XM_006235412.1 | XP_006235474.1 | 2 - 3 | 652 | 102 | ||
GenBank mRNA | BC168699.1 | - | 2 - 3 | 651 | 8362231 | 102 |
DY310142.1 | - | 2 - 3 | 646 | 102 | ||
DY565371.1 | - | 2 - 3 | 422 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |