Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230151.2 | XP_006230213.2 | 11 - 12 | 2311 | 84 | |
GenBank mRNA | AJ295748.1 | - | 8 - 9 | 1817 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |