Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053543.2 | NP_445995.2 | 1 - 2 | 390 | 78 | |
GenBank mRNA | BC087000.1 | - | 1 - 2 | 390 | 7190905 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |