Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021739.2 | NP_068507.2 | 12 - 13 | 1026 | 63 | |
XM_006251170.2 | XP_006251232.1 | 12 - 13 | 1125 | 63 | ||
XM_006251171.2 | XP_006251233.1 | 12 - 13 | 1125 | 63 | ||
XM_006251176.2 | XP_006251238.1 | 12 - 13 | 1106 | 63 | ||
GenBank mRNA | M16112.1 | - | 12 - 13 | 1006 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Rattus norvegicus Probe spans exons |