Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147177.1 | NP_671706.1 | 169 | 77 | ||
XM_006236865.2 | XP_006236927.1 | 243 | 77 | |||
GenBank mRNA | AB001581.1 | - | 156 | 77 | ||
AB002406.1 | - | 169 | 77 | |||
BC072511.1 | - | 154 | 7104777 | 77 | ||
BC086531.1 | - | 187 | 7304521 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |