Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107914.1 | NP_001101384.1 | 2 - 3 | 908 | 88 | |
XM_006238934.1 | XP_006238996.1 | 3 - 4 | 1016 | 88 | ||
XM_006238935.2 | XP_006238997.1 | 3 - 4 | 1379 | 88 | ||
XM_006238936.1 | XP_006238998.1 | 3 - 4 | 1016 | 88 | ||
XM_006238937.1 | XP_006238999.1 | 3 - 4 | 1016 | 88 | ||
XM_006238939.1 | XP_006239001.1 | 3 - 4 | 1016 | 88 | ||
XM_008764141.1 | XP_008762363.1 | 3 - 4 | 1016 | 88 | ||
XM_008764143.1 | XP_008762365.1 | 3 - 4 | 1016 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |