Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127633.1 | NP_001121105.1 | 6 - 7 | 1079 | 84 | |
XM_006238972.2 | XP_006239034.1 | 6 - 7 | 1086 | 84 | ||
GenBank mRNA | BC166486.1 | - | 6 - 7 | 1079 | 7099983 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |