Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031342.1 | NP_112632.1 | 3 - 4 | 195 | 88 | |
GenBank mRNA | AB021645.1 | - | 3 - 4 | 195 | 88 | |
BC070503.1 | - | 3 - 4 | 201 | 7096587 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |