Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 10 - 11 | 1791 | 78 | |
XM_006238646.2 | XP_006238708.1 | 10 - 11 | 1737 | 78 | ||
XM_006238647.2 | XP_006238709.1 | 8 - 9 | 773 | 78 | ||
XM_008763993.1 | XP_008762215.1 | 10 - 11 | 1814 | 78 | ||
GenBank mRNA | BC081913.1 | - | 10 - 11 | 1791 | 7112076 | 78 |
FQ212041.1 | - | 9 - 10 | 1731 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |