Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_573065.5 | XP_573065.3 | 1 - 2 | 594 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |