Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107214.1 | NP_001100684.1 | 3 - 4 | 947 | 78 | |
XM_006250898.2 | XP_006250960.1 | 3 - 4 | 1019 | 78 | ||
XM_006250899.2 | XP_006250961.1 | 3 - 4 | 1018 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |