Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107700.1 | NP_001101170.1 | 5 - 6 | 1418 | 68 | |
GenBank mRNA | BC166708.1 | - | 5 - 6 | 1418 | 5621845 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |