Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014005.1 | NP_001014027.1 | 1 - 2 | 357 | 65 | |
GenBank mRNA | BC083731.1 | - | 1 - 2 | 357 | 7191343 | 65 |
FQ229045.1 | - | 1 - 2 | 428 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |