Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106943.1 | NP_001100413.1 | 1 - 2 | 40 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |