Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766798.1 | XP_008765020.1 | 3 - 4 | 590 | 77 | |
XM_008766799.1 | XP_008765021.1 | 3 - 4 | 533 | 77 | ||
XM_008766800.1 | XP_008765022.1 | 3 - 4 | 323 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |