Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173151.1 | NP_775174.1 | 6 - 7 | 713 | 83 | |
XM_006257016.2 | XP_006257078.1 | 6 - 7 | 674 | 83 | ||
XM_006257017.2 | XP_006257079.1 | 6 - 7 | 1081 | 83 | ||
XM_008773254.1 | XP_008771476.1 | 6 - 7 | 794 | 83 | ||
GenBank mRNA | AF190256.1 | - | 6 - 7 | 713 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |