Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762437.1 | XP_008760659.1 | 11 - 12 | 1591 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |