Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108598.2 | NP_001102068.2 | 2 - 3 | 262 | 61 | |
NM_001244784.1 | NP_001231713.1 | 2 - 3 | 262 | 61 | ||
GenBank mRNA | BC168211.1 | - | 2 - 3 | 91 | 7135378 | 61 |
CB576376.1 | - | 2 - 3 | 139 | 61 | ||
CB585518.1 | - | 2 - 3 | 262 | 61 | ||
CB736780.1 | - | 2 - 3 | 341 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |