Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109107.2 | NP_001102577.2 | 4 - 5 | 774 | 80 | |
GenBank mRNA | BF565650.1 | - | 1 - 2 | 80 | ||
CB316326.1 | - | 2 - 3 | 280 | 80 | ||
CB780985.1 | - | 1 - 2 | 85 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |