Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233702.7 | XP_233702.4 | 10 - 11 | 1909 | 64 | |
GenBank mRNA | BC098821.1 | - | 4 - 5 | 642 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |