Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 15 - 16 | 2350 | 70 | |
GenBank mRNA | AF083341.1 | - | 15 - 16 | 1786 | 70 | |
AF135265.1 | - | 15 - 16 | 2204 | 70 | ||
AY330290.1 | - | 15 - 16 | 1675 | 70 | ||
AY330291.1 | - | 15 - 16 | 1675 | 70 | ||
AY330292.1 | - | 15 - 16 | 1684 | 70 | ||
AY330293.2 | - | 15 - 16 | 1675 | 70 | ||
AY330294.1 | - | 15 - 16 | 1675 | 70 | ||
AY344964.1 | - | 15 - 16 | 1675 | 70 | ||
U40603.1 | - | 15 - 16 | 2350 | 70 | ||
U55995.1 | - | 15 - 16 | 1828 | 70 | ||
U93052.1 | - | 15 - 16 | 1776 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |