Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230017.2 | XP_006230079.1 | 10 - 11 | 1268 | 85 | |
XM_006230018.2 | XP_006230080.1 | 10 - 11 | 1193 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |